5-Bromoisoquinoline ≥98%

Supplier: BeanTown Chemical

220900-1G 220900-250MG 220900-5G
BT220900-1GEA 75 USD
BT220900-1G BT220900-250MG BT220900-5G
5-Bromoisoquinoline ≥98%
5-Bromoisoquinoline

CAS: 34784-04-8; MDL No: MFCD01646405 Solid; Molecular Formula: C9H6BrN ; MW: 208.05 Melting Point: 83-87°


  • Solid

Formula: C₉H₆BrN
MW: 208.06 g/mol
Melting Pt: 83…87 °C
MDL Number: MFCD01646405
CAS Number: 34784-04-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR