Ruthenium(III) iodide, anhydrous
Supplier: BeanTown Chemical
220880-1G
220880-5G
BT220880-1GEA
100
USD
BT220880-1G
BT220880-5G
Ruthenium(III) iodide, anhydrous
Ruthenium(III) iodide
CAS: 13896-65-6; EC No: 237-664-8; MDL No: MFCD00016316
Powder; Linear Formula: RuI3; MW: 481.78
Melting Point: 590° (decomposes)
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
I₃Ru Melting Pt: 590 °C (dec.) |
MDL Number:
MFCD00016316 CAS Number: 13896-65-6 EINECS: 237-664-8 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...