Calcium pyrophosphate ≥96%

Supplier: BeanTown Chemical

220855-1KG 220855-250G
BT220855-1KGEA 72 USD
BT220855-1KG BT220855-250G BT220855-5KG
Calcium pyrophosphate ≥96%
Calcium pyrophosphate

CAS: 7790-76-3; EC No: 232-221-5; MDL No: MFCD00015983 Powder; Linear Formula: Ca2P2O7; MW: 254.10 Melting Point: 1353° Density (g/mL): 3.09


  • Powder

Formula: Ca₂O₇P₂
MW: 254.097 g/mol
Melting Pt: 1353 °C
Density: 3.09 g/mL
MDL Number: MFCD00015983
CAS Number: 7790-76-3
EINECS: 232-221-5

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR