Sodium iodide ≥99.5% ACS

Supplier: BeanTown Chemical

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

220830-100G 220830-500G
BT220830-100GEA 75.82 USD
BT220830-100G BT220830-500G
Sodium iodide ≥99.5% ACS
Sodium iodide

CAS: 7681-82-5; EC No: 231-679-3; MDL No: MFCD00003532; RTECS: WB6475000 Crystalline/Powder; Linear Formula: NaI; MW: 149.89 Melting Point: 661°; Boiling Point: 1300-1304° Density (g/mL): 3.67; Refractive Index: 1.775 Air Sensitive, Hygroscopic, Light Sensitive


  • RTECS: WB6475000
  • Refractive index: 1.775
  • Air Sensitive, Hygroscopic, Light Sensitive
  • Crystalline/Powder

Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1300…1304 °C
Melting Pt: 661 °C
Density: 3.67 g/mL
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 99.5% min.
pH of 5% solution at 25°C 6.0-9.0
Insoluble matter ≤0.01%
Chloride and bromide (Cl, Br) ≤0.01%
Iodate (IO3) ≤3ppm
Phosphate (PO4) ≤0.001%
Sulfate (SO4) ≤0.005%
Barium (Ba) ≤0.002%
Heavy metals (as Pb) ≤5ppm
Iron (Fe) ≤5ppm
Calcium (Ca) ≤0.002%
Magnesium (Mg) ≤0.001%
Potassium (K) ≤0.01%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR