Hafnium ≥99.9% (trace metals basis excluding Zr), pieces, Length 3-12 mm

Supplier: BeanTown Chemical

220820-10G 220820-50G
BT220820-10GEA 111 USD
BT220820-10G BT220820-50G
Hafnium ≥99.9% (trace metals basis excluding Zr), pieces, Length 3-12 mm
Hafnium

CAS: 7440-58-6; EC No: 231-166-4; MDL No: MFCD00011032; RTECS: MG4600000 Solid; Molecular Formula: Hf; MW: 178.49 Melting Point: 2227°; Boiling Point: 4602° Density (g/mL): 13.3


  • RTECS: MG4600000
  • Solid

Formula: Hf
MW: 178.49 g/mol
Boiling Pt: 4602 °C
Melting Pt: 2227 °C
Density: 13.3 g/mL
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR