Polyethylene glycol monomethyl ether, M.W. ∼1900
Supplier: BeanTown Chemical
220805-100G
220805-500G
BT220805-100GEA
43
USD
BT220805-100G
BT220805-500G
Polyethylene glycol monomethyl ether, M.W. ∼1900
Polyethylene glycol monomethyl ether
CAS: 9004-74-4; MDL No: MFCD00084416
Flakes; Linear Formula: CH3(OCH2CH2)nOH; MW ≈2,000
Melting Point: 52-56°; Flash point: 182°C (360°F)
Density (g/mL): 1.094
- Flakes
Formula:
CH₃(OCH₂CH₂)nOH Melting Pt: 52…56 °C Density: 1.094 g/mL Flash Pt: 182 °C (360 °F) |
MDL Number:
MFCD00084416 CAS Number: 9004-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...