2,5-Dibromo-1,4-dimethoxybenzene 98

Supplier: BeanTown Chemical

Synonyms: 1,4-Dibromo-2,5-dimethoxybenzene

220785-10G 220785-50G
BT220785-10GEA 120 USD
BT220785-10G BT220785-50G
2,5-Dibromo-1,4-dimethoxybenzene 98
2,5-Dibromo-1,4-dimethoxybenzene

CAS: 2674-34-2; MDL No: MFCD00025694 Powder; Molecular Formula: C8H8Br2O2; MW: 295.96 Melting Point: 144-149°


  • Powder

Formula: C₈H₈Br₂O₂
MW: 295.96
Melting Pt: 144…149 °C
MDL Number: MFCD00025694
CAS Number: 2674-34-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR