L(+)-Sodium lactate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Sodium (S)-2-hydroxypropanoate, (S)-(+)-2-Hydroxypropanoic acid sodium salt, (S)-(+)-2-Hydroxypropionic acid sodium salt, Sodium-(S)-lactate, (S)-(+)-Lactic acid sodium salt, L(+)-Lactic acid sodium salt
220765-25G
220765-5G
BT220765-25GEA
286.04
USD
BT220765-25G
BT220765-5G
L(+)-Sodium lactate ≥98%
L(+)-Sodium lactate
CAS: 867-56-1; EC No: 212-762-3; MDL No: MFCD00066576
Crystalline; Molecular Formula: C3H5NaO3; MW: 112.06
Melting Point: 163-165°
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
C₃H₅NaO₃ MW: 112.06 g/mol Melting Pt: 163…165 °C |
MDL Number:
MFCD00066576 CAS Number: 867-56-1 EINECS: 212-762-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...