Diisopropylamine dichloroacetate ≥95%

Supplier: BeanTown Chemical

Synonyms: Diisopropylammonium dichloroacetate

220720-1G 220720-250MG 220720-5G
BT220720-1GEA 134 USD
BT220720-1G BT220720-250MG BT220720-5G
Diisopropylamine dichloroacetate ≥95%
Diisopropylamine dichloroacetate

CAS: 660-27-5; EC No: 211-538-2; RTECS: AG6475000 Solid; Molecular Formula: C8H17Cl2NO2; MW: 230.13


  • RTECS: AG6475000
  • Solid

Formula: C₈H₁₇Cl₂NO₂
MW: 230.129 g/mol
MDL Number: MFCD00868288
CAS Number: 660-27-5
EINECS: 211-538-2

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR