Ammonium benzoate ≥99%

Supplier: BeanTown Chemical

Synonyms: Benzoic acid ammonium salt

220665-100G 220665-2KG 220665-500G
BT220665-100GEA 17 USD
BT220665-100G BT220665-2KG BT220665-500G
Ammonium benzoate ≥99%
Ammonium benzoate

CAS: 1863-63-4; EC No: 217-468-9; MDL No: MFCD00013069; RTECS: DG3378000 Crystalline/Solid; Linear Formula: C6H5CO2NH4; Molecular Formula: C7H9NO2; MW: 139.15 Melting Point: 192-198° (decomposes) Moisture Sensitive


  • RTECS: DG3378000
  • Moisture Sensitive
  • Crystalline/Solid

Formula: C₇H₉NO₂
MW: 139.15 g/mol
Melting Pt: 192…198 °C (dec.)
MDL Number: MFCD00013069
CAS Number: 1863-63-4
EINECS: 217-468-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR