1-Benzyl-4-hydroxypiperidine ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-Benzylpiperidin-4-ol

220650-100G 220650-25G 220650-5G
BT220650-100GEA 434 USD
BT220650-100G BT220650-25G BT220650-5G
1-Benzyl-4-hydroxypiperidine ≥97%
1-Benzyl-4-hydroxypiperidine

CAS: 4727-72-4; EC No: 225-226-9; MDL No: MFCD00006503 Solid; Molecular Formula: C12H18NO; MW: 191.27 Melting Point: 61-63°; Boiling Point: 127-128°/2 mmHg Hygroscopic


  • Hygroscopic
  • Solid

Formula: C₁₂H₁₇NO
MW: 191.27 g/mol
Boiling Pt: 127…128 °C/2 mmHg
Melting Pt: 61…63 °C
MDL Number: MFCD00006503
CAS Number: 4727-72-4
EINECS: 225-226-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR