Nicotinamide ≥99%

Supplier: BeanTown Chemical

Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3

220630-1KG 220630-250G
BT220630-1KGEA 89 USD
BT220630-1KG BT220630-250G
Nicotinamide ≥99%
Nicotinamide

CAS: 98-92-0; EC No: 202-713-4; MDL No: MFCD00006395; RTECS: QS3675000 Powder; Molecular Formula: C6H6N2O; MW: 122.13 Melting Point: 128-132°; Flash point: 150°C (302°F) Density (g/mL): 1.4


  • RTECS: QS3675000
  • Powder

Formula: C₆H₆N₂O
MW: 122.13 g/mol
Melting Pt: 128…132 °C
Density: 1.4 g/mL
Flash Pt: 150 °C (302 °F)
MDL Number: MFCD00006395
CAS Number: 98-92-0
EINECS: 202-713-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR