Sodium oleate
Supplier: BeanTown Chemical
220625-100G
220625-25G
220625-500G
BT220625-100GEA
127
USD
BT220625-100G
BT220625-25G
BT220625-500G
Sodium oleate
CAS: 143-19-1; EC No: 205-591-0
Solid; Molecular Formula: C18H33NaO2; MW: 304.45
- Solid
Formula:
C₁₈H₃₃NaO₂ |
MDL Number:
MFCD00004438 CAS Number: 143-19-1 EINECS: 205-591-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...