Iron(II) oxide ≥99.5%
Supplier: BeanTown Chemical
220600-25G
220600-5G
BT220600-25GEA
418.98
USD
BT220600-25G
BT220600-5G
Iron(II) oxide ≥99.5%
Iron(II) oxide
CAS: 1345-25-1; EC No: 215-721-8; MDL No: MFCD00016095
-10 Mesh Powder; Molecular Formula: FeO; MW: 71.85
Density (g/mL): 5.7
Air Sensitive
- Air Sensitive
- -10 Mesh Powder
Formula:
FeO MW: 71.844 g/mol Density: 5.7 g/mL |
MDL Number:
MFCD00016095 CAS Number: 1345-25-1 EINECS: 215-721-8 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...