4-Nitropyrocatechol ≥98%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: 4-Nitrocatechol, 4-Nitrobenzene-1,2-diol

220585-100G 220585-25G 220585-5G
BT220585-100GEA 660 USD
BT220585-100G BT220585-25G BT220585-5G
4-Nitropyrocatechol ≥98%, Technical Grade
4-Nitropyrocatechol

CAS: 3316-09-4; EC No: 222-009-0; MDL No: MFCD00007242 Crystalline; Molecular Formula: C6H5NO4; MW: 155.11 Melting Point: 173-177°


  • Crystalline

Formula: C₆H₅NO₄
MW: 155.11 g/mol
Melting Pt: 173…177 °C
MDL Number: MFCD00007242
CAS Number: 3316-09-4
EINECS: 222-009-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR