4-Nitrobenzaldehyde ≥99%
Supplier: BeanTown Chemical
220570-100G
220570-25G
220570-500G
BT220570-100GEA
126.58
USD
BT220570-100G
BT220570-25G
BT220570-500G
4-Nitrobenzaldehyde ≥99%
4-Nitrobenzaldehyde
CAS: 555-16-8; EC No: 209-084-5; MDL No: MFCD00007346; RTECS: CU7350000
Powder; Molecular Formula: C7H5NO3; MW: 151.12
Melting Point: 103-106°
Air Sensitive
- RTECS: CU7350000
- Air Sensitive
- Powder
Formula:
C₇H₅NO₃ Melting Pt: 103…106 °C |
MDL Number:
MFCD00007346 CAS Number: 555-16-8 EINECS: 209-084-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...