Polyethyleneimine solution, M.W. 50,000-100,000 30% (w/w) in water, branched
Supplier: BeanTown Chemical
Warning
|
Synonyms:
Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)
220550-100G
220550-25G
220550-500G
BT220550-100GEA
167
USD
BT220550-100G
BT220550-25G
BT220550-500G
Polyethyleneimine solution, M.W. 50,000-100,000 30% (w/w) in water, branched
Polyethyleneimine
CAS: 9002-98-6; MDL No: MFCD00803910; RTECS: TQ7915000
Liquid; Linear Formula: (-NHCH2CH2-)x[-N(CH2CH2NH2)CH2CH2-]y
- RTECS: TQ7915000
- Liquid
Formula:
(CH2CH2NH)n |
MDL Number:
MFCD00084427 CAS Number: 9002-98-6 EINECS: 232-399-4 Merck Index: 14,1846 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...