(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt ≥96%
Supplier: BeanTown Chemical
Synonyms:
Methyl N-(Triethylammoniosulphonyl)carbamate, Burgess Reagent
220535-1G
220535-25G
220535-5G
BT220535-1GEA
71
USD
BT220535-1G
BT220535-25G
BT220535-5G
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt ≥96%
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt
CAS: 29684-56-8; MDL No: MFCD00077815
Crystalline/Powder; Molecular Formula: C8H18N2O4S; MW: 238.30
Melting Point: 76-79°
Moisture Sensitive
- Moisture Sensitive
- Crystalline/Powder
Formula:
C₈H₁₈N₂O₄S MW: 238.30 g/mol Melting Pt: 76…79 °C |
MDL Number:
MFCD00077815 CAS Number: 29684-56-8 |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...