1-Bromo-2-iodobenzene ≥98% stabilised
Supplier: BeanTown Chemical
Synonyms:
2-Bromo-1-iodobenzene, 2-Bromoiodobenzene
220530-100G
220530-25G
220530-5G
BT220530-100GEA
411
USD
BT220530-100G
BT220530-25G
BT220530-5G
1-Bromo-2-iodobenzene ≥98% stabilised
1-Bromo-2-iodobenzene
CAS: 583-55-1; EC No: 209-508-9; MDL No: MFCD00001030
Liquid; Molecular Formula: C6H4BrI; MW: 282.90
Melting Point: 9-10°; Boiling Point: 120-121°/15 mmHg; Flash point:
- Refractive index: 1.661
- Light Sensitive
- Liquid
Formula:
C₆H₄BrI Boiling Pt: 120…121 °C/15 mmHg Melting Pt: 9…10 °C Density: 2.203 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00001030 CAS Number: 583-55-1 EINECS: 209-508-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...