Magnesium citrate dibasic hydrate
Supplier: BeanTown Chemical
220525-1KG
220525-250G
220525-5KG
BT220525-1KGEA
264
USD
BT220525-1KG
BT220525-250G
BT220525-5KG
Magnesium citrate dibasic hydrate
Magnesium citrate dibasic hydrate
CAS: 119851-23-9; EC No: 231-923-9; MDL No: MFCD00151706
Powder; Molecular Formula: C6H6MgO7·xH2O; MW: 214.14 (anhydrous)
- Powder
Formula:
C₆H₆MgO₇·xH₂O |
MDL Number:
MFCD00151706 CAS Number: 119851-23-9 EINECS: 231-923-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...