Strontium chloride hexahydrate 99.0-103.0% ACS

Supplier: BeanTown Chemical

220485-100G 220485-500G
BT220485-100GEA 46.67 USD
BT220485-100G BT220485-500G
Strontium chloride hexahydrate 99.0-103.0% ACS
Strontium chloride hexahydrate

CAS: 10025-70-4; EC No: 233-971-6; MDL No: MFCD00149865 Crystalline; Linear Formula: SrCl2·6H2O ; MW: 266.62 Density (g/mL): 1.93; Refractive Index: 1.650 Hygroscopic


  • Refractive index: 1.650
  • Hygroscopic
  • Crystalline

Formula: SrCl₂·6H₂O
MW: 266.62 g/mol
Boiling Pt: 100
Melting Pt: 61
Density: 1.93 g/mL
MDL Number: MFCD00149865
CAS Number: 10025-70-4
EINECS: 233-971-6
Merck Index: 13,08920

Order Now

Specification Test Results

Assay 99.0-103.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR