1-[(4-Methoxyphenyl)ethynyl]-4-n-propylbenzene ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-Methoxy-4-((4-propylphenyl)ethynyl)benzene

220475-1G 220475-5G
BT220475-1GEA 58 USD
BT220475-1G BT220475-5G
1-[(4-Methoxyphenyl)ethynyl]-4-n-propylbenzene ≥99%
1-[(4-Methoxyphenyl)ethynyl]-4-n-propylbenzene

CAS: 39969-26-1; MDL No: MFCD04038795 Crystalline/Powder; Molecular Formula: C18H18O; MW: 250.34 Melting Point: 61°; Boiling Point: 162-164°/1 mmHg


  • Crystalline/Powder

Formula: C₁₈H₁₈O
Boiling Pt: 162…164 °C/1 mmHg
Melting Pt: 61 °C
MDL Number: MFCD04038795
CAS Number: 39969-26-1

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR