(4-Carboxybutyl)triphenylphosphonium bromide ≥98%

Supplier: BeanTown Chemical

Synonyms: (4-Carboxybut-1-yl)(triphenyl)phosphonium bromide

220430-100G 220430-25G 220430-500G
BT220430-100GEA 145 USD
BT220430-100G BT220430-25G BT220430-500G
(4-Carboxybutyl)triphenylphosphonium bromide ≥98%
(4-Carboxybutyl)triphenylphosphonium bromide

CAS: 17814-85-6; EC No: 241-782-5; MDL No: MFCD00011906 Solid; Linear Formula: (C6H5)3P(Br)(CH2)4COOH; Molecular Formula: C23H24BrO2P; MW: 443.31 Melting Point: 204-207° Hygroscopic


  • Hygroscopic
  • Solid

Formula: C₂₃H₂₄BrO₂P
MW: 443.32 g/mol
Melting Pt: 204…207 °C
MDL Number: MFCD00011906
CAS Number: 17814-85-6
EINECS: 241-782-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR