12-Hydroxystearic acid ≥85%, Technical Grade

Supplier: BeanTown Chemical

220410-100G 220410-25G 220410-5G
BT220410-100GEA 607 USD
BT220410-100G BT220410-25G BT220410-5G
12-Hydroxystearic acid ≥85%, Technical Grade
12-Hydroxystearic acid

CAS: 106-14-9; EC No: 203-366-1; MDL No: MFCD00004592; RTECS: WI3700000 Powder/Flakes; Molecular Formula: C18H36O3; MW: 300.48 Melting Point: 74-76°


  • RTECS: WI3700000
  • Powder/Flakes

Formula: C₁₈H₃₆O₃
Melting Pt: 74…76 °C
MDL Number: MFCD00004592
CAS Number: 106-14-9
EINECS: 203-366-1

Order Now

Specification Test Results

Assay 85%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR