Calcium-D-gluconate monohydrate 2.5% (w/w)

Supplier: BeanTown Chemical

220385-30G 220385-60G
BT220385-30GEA 145.72 USD
BT220385-30G BT220385-60G
Calcium-D-gluconate monohydrate 2.5% (w/w)
Calcium-D-gluconate monohydrate

CAS: 299-28-5; MDL No: MFCD00064209 Gel; Molecular Formula: C12H22CaO14·H2O ; MW: 448.39


  • Gel

Formula: C₁₂H₂₂CaO₁₄·H₂O
MW: 448.39 g/mol
Melting Pt: 0 °C (32 °F)
MDL Number: MFCD00150787
CAS Number: 66905-23-5
Merck Index: 14,1670

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR