1-Heptanol ≥99%

Supplier: BeanTown Chemical

Synonyms: Heptyl alcohol

220340-100ML 220340-2.5L 220340-500ML
BT220340-100MLEA 23.6 USD
BT220340-100ML BT220340-2.5L BT220340-500ML
1-Heptanol ≥99%
1-Heptanol

CAS: 111-70-6; EC No: 203-897-9; MDL No: MFCD00002986; RTECS: MK0350000 Liquid; Molecular Formula: C7H16O; MW: 116.20 Melting Point: -36°; Boiling Point: 176°; Flash point: 76°C (169°F) Density (g/mL): 0.822; Refractive Index: 1.424


  • RTECS: MK0350000
  • Refractive index: 1.424
  • Liquid

Formula: H₃C(CH₂)₅CH₂OH
MW: 116.2 g/mol
Boiling Pt: 176 °C
Melting Pt: –36 °C
Density: 0.822 g/mL
Flash Pt: 76 °C (169 °F)
MDL Number: MFCD00002986
CAS Number: 111-70-6
EINECS: 203-897-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR