Tetrapotassium diphosphate, anhydrous ≥96%
Supplier: BeanTown Chemical
Synonyms:
Tetrapotassium diphosphate, Potassium diphosphate, Potassium pyrophosphate, tetra-Potassium pyrophosphate, Potassium pyrophospate
220330-1KG
220330-5KG
BT220330-1KGEA
49
USD
BT220330-1KG
BT220330-5KG
Tetrapotassium diphosphate, anhydrous ≥96%
Tetrapotassium diphosphate
CAS: 7320-34-5; EC No: 230-785-7; MDL No: MFCD00011393
Granular; Linear Formula: K4P2O7; MW: 330.34
Melting Point: 1090°
Density (g/mL): 2.33
Hygroscopic
- Hygroscopic
- Granular
Formula:
K₄O₇P₂ MW: 330.34 g/mol Melting Pt: 1090 °C Density: 2.33 g/mL |
MDL Number:
MFCD00011393 CAS Number: 7320-34-5 EINECS: 230-785-7 |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...