Diphenyl-2-pyridylphosphine ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-(Diphenylphosphino)pyridine
220320-25G
220320-5G
BT220320-25GEA
339
USD
BT220320-25G
BT220320-5G
Diphenyl-2-pyridylphosphine ≥98%
Diphenyl-2-pyridylphosphine
CAS: 37943-90-1; MDL No: MFCD00192108
Solid; Molecular Formula: C17H14NP; MW: 263.27
Melting Point: 82-84°
- Solid
Formula:
C₁₇H₁₄NP Melting Pt: 82…84 °C |
MDL Number:
MFCD00192108 CAS Number: 37943-90-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...