2-Amino-5-chloropyridine ≥97%
Supplier: BeanTown Chemical
220305-100G
220305-25G
220305-500G
BT220305-100GEA
133
USD
BT220305-100G
BT220305-25G
BT220305-500G
2-Amino-5-chloropyridine ≥97%
2-Amino-5-chloropyridine
CAS: 1072-98-6; EC No: 214-020-4; MDL No: MFCD00006324
Crystalline; Molecular Formula: C5H5ClN2 ; MW: 128.56
Melting Point: 135-138°; Boiling Point: 127-128°/11 mmHg; Flash point: 160°C (320°F)
- Crystalline
Formula:
C₅H₅ClN₂ MW: 128.56 g/mol Boiling Pt: 127…128 °C/11 mmHg Melting Pt: 135…138 °C Flash Pt: 160 °C (320 °F) |
MDL Number:
MFCD00006324 CAS Number: 1072-98-6 EINECS: 214-020-4 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...