Malonic acid ≥99%

Supplier: BeanTown Chemical

220300-100G 220300-2.5KG 220300-500G
BT220300-100GEA 47.26 USD
BT220300-100G BT220300-2.5KG BT220300-500G
Malonic acid ≥99%
Malonic acid

CAS: 141-82-2; EC No: 205-503-0; MDL No: MFCD00002707; RTECS: OO0175000 Powder; Linear Formula: CH2(COOH)2; Molecular Formula: C3H4O4; MW: 104.06 Melting Point: 132-135° (decomposes); Flash point: 157°C (314°F) Density (g/mL): 1.6


  • RTECS: OO0175000
  • Powder

Formula: C₃H₄O₄
Melting Pt: 132…135 °C (dec.)
Density: 1.6 g/mL
Flash Pt: 157 °C (314 °F)
MDL Number: MFCD00002707
CAS Number: 141-82-2
EINECS: 205-503-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR