1-Bromo-3,5-bis(trifluoromethyl)benzene ≥98%
Supplier: BeanTown Chemical
Synonyms:
3,5-Bis(trifluoromethyl)-1-bromobenzene, 1-Bromo-3,5-bis(trifluoromethyl)benzene, 5-Bromo-α,α,α,β,β,β-Hexafluoro-m-xylene, 1,3-Bis(trifluoromethyl)-5-bromobenzene, 3,5-Bis(trifluoromethyl)bromobenzene, 5-Bromo-α,α,α,α',α',α'-Hexafluoro-m-xylene
220285-10G
220285-250G
220285-50G
BT220285-10GEA
35
USD
BT220285-10G
BT220285-250G
BT220285-50G
1-Bromo-3,5-bis(trifluoromethyl)benzene ≥98%
1-Bromo-3,5-bis(trifluoromethyl)benzene
CAS: 328-70-1; EC No: 206-334-5; MDL No: MFCD00000381
Liquid; Molecular Formula: C8H3BrF6; MW: 293.00
Melting Point: -16°; Boiling Point: 154°; Flash point: 112°C (234°F)
Density (g/mL): 1.699; Refractive Index: 1.427
- Refractive index: 1.427
- Liquid
Formula:
C₈H₃BrF₆ Boiling Pt: 154 °C Melting Pt: –16 °C Density: 1.699 g/mL Flash Pt: 112 °C (234 °F) |
MDL Number:
MFCD00000381 CAS Number: 328-70-1 EINECS: 206-334-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...