1,3-Dibromo-2,2-dimethoxypropane ≥99%

Supplier: BeanTown Chemical

220250-100G 220250-25G 220250-5G
BT220250-100GEA 1120 USD
BT220250-100G BT220250-25G BT220250-5G
1,3-Dibromo-2,2-dimethoxypropane ≥99%
1,3-Dibromo-2,2-dimethoxypropane

CAS: 22094-18-4; EC No: 000-000-0; MDL No: MFCD00051792 Solid; Molecular Formula: C5H10Br2O2; MW: 261.94


  • Solid

Formula: BrH₂CC(OCH₃)₂CH₂Br
MW: 261.94 g/mol
MDL Number: MFCD00051792
CAS Number: 22094-18-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR