Chrysin ≥98%

Supplier: BeanTown Chemical

Synonyms: 5,7-Dihydroxy-2-phenyl-4H-chromen-4-one, 5,7-Dihydroxyflavone, 5,7-Dihydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 5,7-Dihydroxy-2-phenyl-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-phenyl-4-benzopyrone, 5,7-Dihydroxy-2-phenylchromen-4-one

220235-25G 220235-5G
BT220235-25GEA 95 USD
BT220235-25G BT220235-5G
Chrysin ≥98%
Chrysin

CAS: 480-40-0; EC No: 207-549-7; MDL No: MFCD00006834; RTECS: LK8329050 Powder; Molecular Formula: C15H10O4; MW: 254.24 Melting Point: 284-286°


  • RTECS: LK8329050
  • Powder

Formula: C₁₅H₁₀O₄
MW: 254.24 g/mol
Boiling Pt: 429
Melting Pt: 284…286 °C
MDL Number: MFCD00006834
CAS Number: 480-40-0
EINECS: 207-549-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR