Dithiooxamide ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethanebis(thioamide), Rubeanic acid

220220-25G 220220-5G
BT220220-25GEA 355 USD
BT220220-25G BT220220-5G
Dithiooxamide ≥98%
Dithiooxamide

CAS: 79-40-3; EC No: 201-203-9; MDL No: MFCD00004941; RTECS: RP1575000 Powder; Linear Formula: NH2SCSSNH2; Molecular Formula: C2H4N2S2; MW: 120.20 Melting Point: 300°


  • RTECS: RP1575000
  • Powder

Formula: C₂H₄N₂S₂
Melting Pt: >300 °C
MDL Number: MFCD00004941
CAS Number: 79-40-3
EINECS: 201-203-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR