Tetrakis(triphenylphosphine)platinum(0)

Supplier: BeanTown Chemical

220205-1G 220205-5G
BT220205-1GEA 134 USD
BT220205-1G BT220205-5G
Tetrakis(triphenylphosphine)platinum(0)
Tetrakis(triphenylphosphine)platinum(0)

CAS: 14221-02-4; EC No: 238-087-4; MDL No: MFCD00010014 Crystalline/Powder; Linear Formula: Pt[P(C6H5)3]4; Molecular Formula: C72H60P4Pt; MW: 1244.21 Air Sensitive, Light Sensitive


  • Air Sensitive, Light Sensitive
  • Crystalline/Powder

Formula: C₇₂H₆₀P₄Pt
MDL Number: MFCD00010014
CAS Number: 14221-02-4
EINECS: 238-087-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR