Aluminium potassium sulphate dodecahydrate 98.0-102.0% ACS

Supplier: BeanTown Chemical

Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium

220190-2KG 220190-500G
BT220190-2KGEA 135 USD
BT220190-2KG BT220190-500G
Aluminium potassium sulphate dodecahydrate 98.0-102.0% ACS
Aluminium potassium sulphate dodecahydrate

CAS: 7784-24-9; EC No: 233-141-3; MDL No: MFCD00149143; RTECS: WS5690000 Granular; Linear Formula: KAl(SO4)2·12H2O ; MW: 474.39 Melting Point: 92° Density (g/mL): 1.757


  • RTECS: WS5690000
  • Granular

Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Melting Pt: 92.5
Density: 1.757 g/mL
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Order Now

Specification Test Results

Assay 98.0-102.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR