Tetramethylurea ≥99%
Supplier: BeanTown Chemical
CAS: 632-22-4; EC No: 211-173-9; MDL No: MFCD00008319; RTECS: YU2625000
Liquid; Linear Formula: (CH3)2NCON(CH3)2; Molecular Formula: C5H12N2O; MW: 116.16
Melting Point: -1°; Boiling Point: 176-178°; Flash point: 75°C (167°F)
Density (g/mL): 0.968; Refractive Index: 1.451
Hygroscopic
- RTECS: YU2625000
- Refractive index: 1.451
- Hygroscopic
- Liquid
Formula:
C₅H₁₂N₂O Boiling Pt: 176…178 °C Melting Pt: –1 °C Density: 0.968 g/mL Flash Pt: 75 °C (167 °F) |
MDL Number:
MFCD00008319 CAS Number: 632-22-4 EINECS: 211-173-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...