Potassium acetate ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Acetic acid potassium salt

220160-100G 220160-2KG 220160-500G
BT220160-100GEA 32.86 USD
BT220160-100G BT220160-2KG BT220160-500G
Potassium acetate ≥99.0% ACS
Potassium acetate

CAS: 127-08-2; EC No: 204-822-2; MDL No: MFCD00012458; RTECS: AJ3325000 Crystalline; Linear Formula: CH3COOK; MW: 98.15 Melting Point: 292° Density (g/mL): 1.57 Hygroscopic


  • RTECS: AJ3325000
  • Hygroscopic
  • Crystalline

Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304
Density: 1.57 g/mL
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR