2-Fluoro-5-nitrobenzaldehyde ≥98%
Supplier: BeanTown Chemical
220140-1G
220140-5G
BT220140-1GEA
80
USD
BT220140-1G
BT220140-5G
2-Fluoro-5-nitrobenzaldehyde ≥98%
2-Fluoro-5-nitrobenzaldehyde
CAS: 27996-87-8; MDL No: MFCD00042298
Powder; Molecular Formula: C7H4FNO3; MW: 169.11
Melting Point: 57-60°
Air Sensitive
- Air Sensitive
- Powder
Formula:
C₇H₄FNO₃ MW: 169.11 g/mol Melting Pt: 57…60 °C |
MDL Number:
MFCD00042298 CAS Number: 27996-87-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...