5-Methoxy-2-methylindole ≥99%

Supplier: BeanTown Chemical

Synonyms: 5-Methoxy-2-methyl-1H-indole

220130-1G 220130-25G 220130-5G
BT220130-1GEA 36.24 USD
BT220130-1G BT220130-25G BT220130-5G
5-Methoxy-2-methylindole ≥99%
5-Methoxy-2-methylindole

CAS: 1076-74-0; EC No: 214-066-5; MDL No: MFCD00005620 Solid; Molecular Formula: C10H11NO; MW: 161.20 Melting Point: 86-88°


  • Solid

Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD00005620
CAS Number: 1076-74-0
EINECS: 214-066-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR