DL-Sodium lactate 60% (w/w) in water
Supplier: BeanTown Chemical
Synonyms:
(±)-Lactic acid sodium salt, (±)-2-Hydroxypropionic acid sodium salt, Lactate sodium, DL-Lactic acid sodium salt, (±)-2-Hydroxypropanoic acid sodium salt, Sodium 2-hydroxypropanoate
220110-1L
220110-250ML
BT220110-1LEA
127.32
USD
BT220110-1L
BT220110-250ML
DL-Sodium lactate 60% (w/w) in water
DL-Sodium lactate
CAS: 72-17-3; EC No: 200-772-0; MDL No: MFCD00065400; RTECS: OD5680000
Liquid; Linear Formula: CH3CH(OH)CO2Na ; Molecular Formula: C3H5NaO3; MW: 112.06
Density (g/mL): 1.326
- RTECS: OD5680000
- Liquid
Formula:
H₃CCH(OH)COONa MW: 112.06 g/mol Density: 1.326 g/mL |
MDL Number:
MFCD00065400 CAS Number: 72-17-3 EINECS: 200-772-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...