1,2-Diphenylethane ≥99%

Supplier: BeanTown Chemical

Synonyms: Dibenzyl, Bibenzyl

220095-100G 220095-25G 220095-500G
BT220095-100GEA 159 USD
BT220095-100G BT220095-25G BT220095-500G
1,2-Diphenylethane ≥99%
1,2-Diphenylethane

CAS: 103-29-7; EC No: 203-096-4; MDL No: MFCD00004796; RTECS: DT4375000 Powder; Linear Formula: C6H5CH2CH2C6H5; Molecular Formula: C14H14; MW: 182.26 Melting Point: 50-53°; Boiling Point: 284°; Flash point: 110°C (230°F) Density (g/mL): 1.014


  • RTECS: DT4375000
  • Powder

Formula: (C₆H₅)CH₂CH₂(C₆H₅)
MW: 182.27 g/mol
Boiling Pt: 284 °C
Melting Pt: 50…53 °C
Density: 1.014 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00004796
CAS Number: 103-29-7
EINECS: 203-096-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR