Carbonylbis(triphenylphosphine)rhodium(I) chloride
Supplier: BeanTown Chemical
Synonyms:
Carbonylchlorobis(triphenylphosphine)rhodium(I), Bis(triphenylphosphine)carbonylrhodium(I) Chloride, Chlorocarbonylbis(triphenylphosphine)rhodium(I)
220085-1G
220085-250MG
220085-5G
BT220085-1GEA
240
USD
BT220085-1G
BT220085-250MG
BT220085-5G
Carbonylbis(triphenylphosphine)rhodium(I) chloride
Carbonylbis(triphenylphosphine)rhodium(I) chloride
CAS: 13938-94-8; EC No: 237-712-8; MDL No: MFCD00044273
Crystalline; Linear Formula: RhCl(CO)[P(C6H5)3]2; Molecular Formula: C37H30ClOP2Rh; MW: 690.94
Melting Point: 224-227° (decomposes)
- Crystalline
Formula:
C₃₇H₃₀ClOP₂Rh Melting Pt: 224…227 °C (dec.) |
MDL Number:
MFCD00044273 CAS Number: 13938-94-8 EINECS: 237-712-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...