Disodium malonate ≥99%
Supplier: BeanTown Chemical
220050-100G
220050-25G
BT220050-100GEA
86.28
USD
BT220050-100G
BT220050-25G
Disodium malonate ≥99%
Disodium malonate
CAS: 141-95-7; EC No: 205-514-0; MDL No: MFCD00002708; RTECS: OO1750000
Crystalline/Powder; Linear Formula: CH2(COONa)2; Molecular Formula: C3H2Na2O4; MW: 148.03
- RTECS: OO1750000
- Crystalline/Powder
Formula:
C₃H₂Na₂O₄ |
MDL Number:
MFCD00002708 CAS Number: 141-95-7 EINECS: 205-514-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...