Polyethylene glycol monomethyl ether, M.W. ∼750
Supplier: BeanTown Chemical
220030-100G
220030-500G
BT220030-100GEA
27.01
USD
BT220030-100G
BT220030-500G
Polyethylene glycol monomethyl ether, M.W. ∼750
Polyethylene glycol monomethyl ether
CAS: 9004-74-4; MDL No: MFCD00084416
Semisolid/Liquid; Linear Formula: CH3(OCH2CH2)nOH; MW ≈750
Melting Point: 30°; Flash point: 182°C (360°F)
Density (g/mL): 1.094; Refractive Index: 1.459
- Refractive index: 1.459
- Semisolid/Liquid
Formula:
CH₃(OCH₂CH₂)nOH Melting Pt: 30 °C Density: 1.094 g/mL Flash Pt: 182 °C (360 °F) |
MDL Number:
MFCD00084416 CAS Number: 9004-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...