Ammonium tetrachlorocuprate dihydrate

Supplier: BeanTown Chemical

220015-100G 220015-500G
BT220015-100GEA 86.19 USD
BT220015-100G BT220015-500G
Ammonium tetrachlorocuprate dihydrate
Ammonium tetrachlorocuprate dihydrate

CAS: 10060-13-6; EC No: 239-690-5; MDL No: MFCD00150384 Crystalline; Linear Formula: (NH4)2CuCl4·2H2O; MW: 277.47


  • Crystalline

Formula: H₈Cl₄CuN₂·2H₂O
MDL Number: MFCD00150384
CAS Number: 10060-13-6
EINECS: 239-690-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR