Diammonium tetrachloropalladate
Supplier: BeanTown Chemical
220005-1G
220005-5G
BT220005-1GEA
69
USD
BT220005-1G
BT220005-5G
Diammonium tetrachloropalladate
Diammonium tetrachloropalladate
CAS: 13820-40-1; EC No: 237-498-6; MDL No: MFCD00010884; RTECS: BP5400000
Powder; Linear Formula: (NH4)2PdCl4; MW: 284.31
Density (g/mL): 2.17
Hygroscopic
- RTECS: BP5400000
- Hygroscopic
- Powder
Formula:
H₈Cl₄N₂Pd Density: 2.17 g/mL |
MDL Number:
MFCD00010884 CAS Number: 13820-40-1 EINECS: 237-498-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...