(S)-(+)-2-Aminobutyric acid ≥98%
Supplier: BeanTown Chemical
219995-1G
219995-25G
219995-5G
BT219995-1GEA
40
USD
BT219995-1G
BT219995-25G
BT219995-5G
(S)-(+)-2-Aminobutyric acid ≥98%
(S)-(+)-2-Aminobutyric acid
CAS: 1492-24-6; EC No: 216-083-3; MDL No: MFCD00064415
Crystalline/Solid; Molecular Formula: C4H9NO2; MW: 103.12
- Crystalline/Solid
Formula:
C₄H₉NO₂ MW: 103.12 g/mol |
MDL Number:
MFCD00064415 CAS Number: 1492-24-6 EINECS: 216-083-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...