Potassium dihydrogen citrate hydrate ≥99% (dry weight)
Supplier: BeanTown Chemical
Synonyms:
Potassium citrate monobasic hydrate, Citric acid monopotassium salt hydrate, mono-Potassium citrate hydrate
219990-2KG
219990-500G
BT219990-2KGEA
312.55
USD
BT219990-2KG
BT219990-500G
Potassium dihydrogen citrate hydrate ≥99% (dry weight)
Potassium dihydrogen citrate hydrate
CAS: 866-83-1; EC No: 212-753-4; MDL No: MFCD00036472
Crystalline; Linear Formula: COOHCH2C(OH)(COOH)CH2COOK·xH2O; Molecular Formula: C6H7KO7·xH2O; MW: 230.21 (anhydrous)
Hygroscopic
- Crystalline
- Hygroscopic
Formula:
C₆H₇KO₇·xH₂O MW: 230.22 g/mol (anhy) |
MDL Number:
MFCD00036472 CAS Number: 866-83-1 EINECS: 212-753-4 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...