Methyl-2-amino-4,5-dimethoxybenzoate ≥98%

Supplier: BeanTown Chemical

Synonyms: Methyl 4,5-dimethoxyanthranilate, 2-Amino-4,5-dimethoxy benzoate, Methyl 2-amino-4,5-dimethoxybenzoate, 2-Amino-4,5-dimethoxybenzoic acid methyl ester

219975-10G 219975-50G
BT219975-10GEA 52 USD
BT219975-10G BT219975-50G
Methyl-2-amino-4,5-dimethoxybenzoate ≥98%
Methyl-2-amino-4,5-dimethoxybenzoate

CAS: 26759-46-6; EC No: 247-967-7; MDL No: MFCD00014904 Powder; Molecular Formula: C10H13NO4; MW: 211.22 Melting Point: 128-133°


  • Powder

Formula: C₁₀H₁₃NO₄
MW: 211.22 g/mol
Melting Pt: 128…133 °C
MDL Number: MFCD00014904
CAS Number: 26759-46-6
EINECS: 247-967-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR